Assay Details
| Gene Symbol: INSL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174365.2 | NP_776790.1 | 1 - 2 | 228 | 97 | |
| GenBank mRNA | AF094580.1 | - | 1 - 2 | 228 | 97 | |
| BC102547.1 | - | 1 - 2 | 210 | 97 | ||
More Information
Set Membership: |
|

INSL3
Genomic Map