Assay Details
| Gene Symbol: ACVR2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174495.2 | NP_776920.1 | 7 - 8 | 1117 | 71 | |
| GenBank mRNA | HQ703505.1 | - | 1 - 2 | 147 | 71 | |
| U57707.1 | - | 7 - 8 | 1117 | 71 | ||
More Information
Set Membership: |
|

ACVR2B
Genomic Map