Assay Details
| Gene Symbol: GABRA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174543.2 | NP_776968.1 | 1 - 2 | 114 | 80 | |
| GenBank mRNA | BC123424.1 | - | 1 - 2 | 243 | 80 | |
| X61456.1 | - | 1 | 114 | 80 | ||
More Information
Set Membership: |
|

GABRA4
Genomic Map