Assay Details
| Gene Symbol: UQCRC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174629.2 | NP_777054.1 | 3 - 4 | 305 | 59 | |
| GenBank mRNA | BC104500.2 | - | 3 - 4 | 291 | 59 | |
| X59692.1 | - | 305 | 59 | |||
More Information
Set Membership: |
|

UQCRC1
Genomic Map