Assay Details
| Gene Symbol: UQCRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001370607.1 | NP_001357536.1 | 6 - 7 | 634 | 58 | |
| GenBank mRNA | BC102337.1 | - | 6 - 7 | 603 | 58 | |
| BT020993.1 | - | 6 - 7 | 586 | 58 | ||
| X59693.1 | - | 592 | 58 | |||
More Information
Set Membership: |
|

UQCRC2
Genomic Map