Assay Details
| Gene Symbol: CLEC3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174633.2 | NP_777058.1 | 1 - 2 | 237 | 81 | |
| GenBank mRNA | U22298.1 | - | 1 - 2 | 237 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CLEC3A| Gene Symbol: CLEC3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174633.2 | NP_777058.1 | 1 - 2 | 237 | 81 | |
| GenBank mRNA | U22298.1 | - | 1 - 2 | 237 | 81 | |
Set Membership: |
|