Assay Details
| Gene Symbol: GUCY1B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174641.1 | NP_777066.1 | 9 - 10 | 1257 | 66 | |
| GenBank mRNA | BC133308.1 | - | 9 - 10 | 1254 | 66 | |
| Y00770.1 | - | 1257 | 66 | |||
More Information
Set Membership: |
|

GUCY1B1
Genomic Map