Assay Details
| Gene Symbol: PPP1R1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174647.2 | NP_777072.1 | 4 - 5 | 583 | 71 | |
| GenBank mRNA | BC118264.1 | - | 4 - 5 | 318 | 71 | |
| M27444.1 | - | 4 - 5 | 583 | 71 | ||
More Information
Set Membership: |
|

PPP1R1B
Genomic Map