Assay Details
| Gene Symbol: TNFRSF1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174674.2 | NP_777099.1 | 5 - 6 | 842 | 60 | |
| GenBank mRNA | BC113278.1 | - | 5 - 6 | 658 | 60 | |
| U90937.1 | - | 5 - 6 | 842 | 60 | ||
More Information
Set Membership: |
|

TNFRSF1A
Genomic Map