Assay Details
| Gene Symbol: ATP1B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174677.2 | NP_777102.1 | 2 - 3 | 258 | 69 | |
| GenBank mRNA | BC148007.1 | - | 2 - 3 | 678 | 69 | |
| U45944.1 | - | 2 - 3 | 258 | 69 | ||
More Information
Set Membership: |
|

ATP1B2
Genomic Map