Assay Details
| Gene Symbol: FDXR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174691.1 | NP_777116.1 | 6 - 7 | 666 | 64 | |
| GenBank mRNA | BC118190.1 | - | 6 - 7 | 651 | 64 | |
| BT021762.1 | - | 6 - 7 | 649 | 64 | ||
| BT021817.1 | - | 6 - 7 | 651 | 64 | ||
| D00211.1 | - | 645 | 64 | |||
| M17029.1 | - | 6 - 7 | 666 | 64 | ||
| X13736.1 | - | 151 | 64 | |||
More Information
Set Membership: |
|

FDXR
Genomic Map