Assay Details
| Gene Symbol: CELA2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174714.2 | NP_777139.1 | 3 - 4 | 238 | 110 | |
| GenBank mRNA | BC134791.1 | - | 3 - 4 | 251 | 110 | |
| X97635.1 | - | 238 | 110 | |||
More Information
Set Membership: |
|

CELA2A
Genomic Map