Assay Details
| Gene Symbol: GUCA1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174786.1 | NP_777211.1 | 1 - 2 | 317 | 61 | |
| GenBank mRNA | L43001.1 | - | 1 - 2 | 317 | 61 | |
| U32856.1 | - | 1 - 2 | 218 | 61 | ||
More Information
Set Membership: |
|

GUCA1B
Genomic Map