Assay Details
| Gene Symbol: CRYBB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174807.1 | NP_777232.1 | 5 - 6 | 506 | 101 | |
| GenBank mRNA | M22466.1 | - | 5 - 6 | 506 | 101 | |
More Information
Set Membership: |
|

CRYBB2
Genomic Map