Assay Details
| Gene Symbol: GLYCAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174828.2 | NP_777253.1 | 1 - 2 | 126 | 59 | |
| GenBank mRNA | BC108244.1 | - | 1 - 2 | 127 | 59 | |
| D26176.1 | - | 1 - 2 | 109 | 59 | ||
| L36854.1 | - | 1 - 2 | 126 | 59 | ||
| LC534834.1 | - | 1 - 2 | 108 | 59 | ||
| X79406.1 | - | 126 | 59 | |||
More Information
Set Membership: |
|

GLYCAM1
Genomic Map