Assay Details
| Gene Symbol: PCTP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174835.2 | NP_777260.1 | 4 - 5 | 565 | 65 | |
| GenBank mRNA | BC116120.1 | - | 4 - 5 | 568 | 65 | |
| U21660.1 | - | 4 - 5 | 565 | 65 | ||
More Information
Set Membership: |
|

PCTP
Genomic Map