Assay Details
| Gene Symbol: SMN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175701.1 | NP_783632.1 | 2 - 3 | 159 | 122 | |
| GenBank mRNA | AF035323.1 | - | 2 - 3 | 159 | 122 | |
| AF466193.1 | - | 2 - 3 | 159 | 122 | ||
| AF466194.1 | - | 2 - 3 | 168 | 122 | ||
| BC148903.1 | - | 2 - 3 | 184 | 122 | ||
More Information
Set Membership: |
|

SMN2
Genomic Map