Assay Details
| Gene Symbol: SMN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175701.1 | NP_783632.1 | 7 - 8 | 837 | 129 | |
| GenBank mRNA | AF035323.1 | - | 7 - 8 | 837 | 129 | |
More Information
Set Membership: |
|

SMN2
Genomic Map