Assay Details
| Gene Symbol: NDUFS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175800.2 | NP_786994.1 | 3 - 4 | 376 | 107 | |
| GenBank mRNA | BC102292.1 | - | 3 - 4 | 377 | 107 | |
| X63215.1 | - | 376 | 107 | |||
More Information
Set Membership: |
|

NDUFS4
Genomic Map