Assay Details
| Gene Symbol: NDUFB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175822.2 | NP_787016.1 | 2 - 3 | 350 | 132 | |
| GenBank mRNA | AB098753.1 | - | 1 - 2 | 122 | 132 | |
| BC103187.1 | - | 2 - 3 | 326 | 132 | ||
| X64898.1 | - | 350 | 132 | |||
More Information
Set Membership: |
|

NDUFB4
Genomic Map