Assay Details
| Gene Symbol: NDUFV3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175830.1 | NP_787024.1 | 1 - 2 | 78 | 60 | |
| GenBank mRNA | BC103274.1 | - | 1 - 2 | 69 | 60 | |
| X59048.1 | - | 78 | 60 | |||
More Information
Set Membership: |
|

NDUFV3
Genomic Map