Assay Details
| Gene Symbol: ATP5MC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176613.2 | NP_788786.1 | 1 - 2 | 59 | 84 | |
| GenBank mRNA | AB098947.1 | - | 1 - 2 | 75 | 84 | |
| AB099013.1 | - | 1 - 2 | 79 | 84 | ||
| BC111613.1 | - | 2 - 3 | 94 | 84 | ||
| X05219.1 | - | 59 | 84 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC111613.1, NM_176613.2, X05219.1 transcripts.
Set Membership: |
|

ATP5MC2
Genomic Map