Assay Details
| Gene Symbol: ATP5MC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176613.2 | NP_788786.1 | 2 - 3 | 148 | 93 | |
| GenBank mRNA | AB098947.1 | - | 2 - 3 | 164 | 93 | |
| AB099013.1 | - | 2 - 3 | 168 | 93 | ||
| BC111613.1 | - | 3 - 4 | 183 | 93 | ||
| X05219.1 | - | 148 | 93 | |||
More Information
Set Membership: |
|

ATP5MC2
Genomic Map