Assay Details
| Gene Symbol: MTCH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317336.2 | NP_001304265.2 | 5 - 6 | 513 | 62 | |
| NM_176634.4 | NP_788807.3 | 5 - 6 | 513 | 62 | ||
| GenBank mRNA | AB042815.1 | - | 5 - 6 | 419 | 62 | |
| BC150080.1 | - | 5 - 6 | 513 | 62 | ||
| BT020931.1 | - | 5 - 6 | 448 | 62 | ||
More Information
Set Membership: |
|

MTCH2
Genomic Map