Assay Details
| Gene Symbol: NDUFS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176652.2 | NP_788825.1 | 2 - 3 | 256 | 95 | |
| GenBank mRNA | BC102919.1 | - | 2 - 3 | 321 | 95 | |
| X63220.1 | - | 256 | 95 | |||
More Information
Set Membership: |
|

NDUFS5
Genomic Map