Assay Details
| Gene Symbol: FCGRT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176657.1 | NP_788830.1 | 6 - 7 | 1102 | 103 | |
| GenBank mRNA | AF139106.1 | - | 6 - 7 | 1102 | 103 | |
| AF221522.1 | - | 6 - 7 | 1034 | 103 | ||
| BC102159.1 | - | 6 - 7 | 1140 | 103 | ||
More Information
Set Membership: |
|

FCGRT
Genomic Map