Assay Details
| Gene Symbol: NDUFA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176659.2 | NP_788832.1 | 3 - 4 | 174 | 61 | |
| GenBank mRNA | BC102727.1 | - | 3 - 4 | 206 | 61 | |
| X63217.1 | - | 174 | 61 | |||
More Information
Set Membership: |
|

NDUFA3
Genomic Map