Assay Details
| Gene Symbol: COX7A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176674.2 | NP_788847.1 | 3 - 4 | 247 | 78 | |
| GenBank mRNA | BC114907.1 | - | 3 - 4 | 225 | 78 | |
| X56739.1 | - | 247 | 78 | |||
More Information
Set Membership: |
|

COX7A1
Genomic Map