Assay Details
| Gene Symbol: LRAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177503.2 | NP_803469.1 | 539 | 105 | ||
| GenBank mRNA | AF275344.1 | - | 539 | 105 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LRAT| Gene Symbol: LRAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177503.2 | NP_803469.1 | 539 | 105 | ||
| GenBank mRNA | AF275344.1 | - | 539 | 105 | ||
Set Membership: |
|