Assay Details
| Gene Symbol: QPCT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177506.2 | NP_803472.1 | 3 - 4 | 632 | 64 | |
| GenBank mRNA | BC120315.1 | - | 3 - 4 | 551 | 64 | |
| M80626.1 | - | 3 - 4 | 632 | 64 | ||
More Information
Set Membership: |
|

QPCT
Genomic Map