Assay Details
| Gene Symbol: UMPS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177508.1 | NP_803474.1 | 3 - 4 | 1016 | 82 | |
| GenBank mRNA | BC112872.1 | - | 3 - 4 | 1014 | 82 | |
| X65125.1 | - | 1016 | 82 | |||
More Information
Set Membership: |
|

UMPS
Genomic Map