Assay Details
| Gene Symbol: GNRHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177514.2 | NP_803480.1 | 1 - 2 | 600 | 102 | |
| GenBank mRNA | U00934.1 | - | 1 - 2 | 600 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GNRHR| Gene Symbol: GNRHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177514.2 | NP_803480.1 | 1 - 2 | 600 | 102 | |
| GenBank mRNA | U00934.1 | - | 1 - 2 | 600 | 102 | |
Set Membership: |
|