Assay Details
| Gene Symbol: GALNT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177519.3 | NP_803485.1 | 2 - 3 | 339 | 88 | |
| GenBank mRNA | L07780.1 | - | 2 - 3 | 373 | 88 | |
| L17437.1 | - | 1 - 2 | 201 | 88 | ||
More Information
Set Membership: |
|

GALNT1
Genomic Map