Assay Details
| Gene Symbol: SULT1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177521.2 | NP_803487.1 | 1 - 2 | 187 | 72 | |
| GenBank mRNA | BC102274.1 | - | 3 - 4 | 508 | 72 | |
| U35253.1 | - | 1 - 2 | 187 | 72 | ||
More Information
Set Membership: |
|

SULT1A1
Genomic Map