Assay Details
| Gene Symbol: SULT1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177521.2 | NP_803487.1 | 5 - 6 | 637 | 80 | |
| GenBank mRNA | U35253.1 | - | 5 - 6 | 637 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SULT1A1| Gene Symbol: SULT1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177521.2 | NP_803487.1 | 5 - 6 | 637 | 80 | |
| GenBank mRNA | U35253.1 | - | 5 - 6 | 637 | 80 | |
Set Membership: |
|