Assay Details
| Gene Symbol: DBH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180995.2 | NP_851338.1 | 8 - 9 | 1366 | 64 | |
| GenBank mRNA | AF118638.1 | - | 8 - 9 | 1357 | 64 | |
| BT026311.1 | - | 8 - 9 | 1352 | 64 | ||
| J02890.1 | - | 8 - 9 | 1366 | 64 | ||
| J02909.1 | - | 8 - 9 | 1245 | 64 | ||
| J05160.1 | - | 8 - 9 | 1350 | 64 | ||
More Information
Set Membership: |
|

DBH
Genomic Map