Assay Details
| Gene Symbol: CHGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181005.2 | NP_851348.1 | 3 - 4 | 378 | 63 | |
| GenBank mRNA | BC105515.1 | - | 3 - 4 | 365 | 63 | |
| M16971.1 | - | 3 - 4 | 317 | 63 | ||
| U73523.1 | - | 3 - 4 | 378 | 63 | ||
| X04012.1 | - | 370 | 63 | |||
| X04298.1 | - | 330 | 63 | |||
More Information
Set Membership: |
|

CHGA
Genomic Map