Assay Details
| Gene Symbol: SFTPD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181026.2 | NP_851369.1 | 7 - 8 | 841 | 89 | |
| GenBank mRNA | X75911.1 | - | 841 | 89 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SFTPD| Gene Symbol: SFTPD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181026.2 | NP_851369.1 | 7 - 8 | 841 | 89 | |
| GenBank mRNA | X75911.1 | - | 841 | 89 | ||
Set Membership: |
|