Assay Details
| Gene Symbol: PTGER3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181032.1 | NP_851375.1 | 2 - 3 | 1070 | 84 | |
| GenBank mRNA | D21345.1 | - | 2 - 3 | 1070 | 84 | |
More Information
Set Membership: |
|

PTGER3
Genomic Map