Assay Details
| Gene Symbol: PLP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203363.1 | NP_976239.1 | 3 - 4 | 434 | 53 | |
| GenBank mRNA | AY192437.1 | - | 3 - 4 | 434 | 53 | |
| BC103120.1 | - | 3 - 4 | 454 | 53 | ||
More Information
Set Membership: |
|

PLP2
Genomic Map