Assay Details
| Gene Symbol: PLP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203363.1 | NP_976239.1 | 4 - 5 | 520 | 56 | |
| GenBank mRNA | AY192437.1 | - | 4 - 5 | 520 | 56 | |
| BC103120.1 | - | 4 - 5 | 540 | 56 | ||
More Information
Set Membership: |
|

PLP2
Genomic Map