Assay Details
| Gene Symbol: LAPTM4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205799.1 | NP_991368.1 | 5 - 6 | 650 | 96 | |
| GenBank mRNA | AB098971.1 | - | 3 - 4 | 296 | 96 | |
| AY528247.1 | - | 5 - 6 | 650 | 96 | ||
| BC105519.1 | - | 5 - 6 | 625 | 96 | ||
More Information
Set Membership: |
|

LAPTM4A
Genomic Map