Assay Details
| Gene Symbol: HSD11B1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206969.1 | NP_996852.1 | 5 - 6 | 532 | 65 | |
| GenBank mRNA | AY554049.1 | - | 5 - 6 | 532 | 65 | |
| BC149481.1 | - | 3 - 4 | 704 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC149481.1 transcripts.
Set Membership: |
|

HSD11B1L
Genomic Map