Assay Details
| Gene Symbol: UPK3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012672.1 | NP_001012690.1 | 4 - 5 | 696 | 60 | |
| GenBank mRNA | AY233461.1 | - | 4 - 5 | 696 | 60 | |
More Information
Set Membership: |
|

UPK3B
Genomic Map