Assay Details
| Gene Symbol: COX6B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012684.1 | NP_001012702.1 | 2 - 3 | 423 | 77 | |
| GenBank mRNA | AY152401.1 | - | 2 - 3 | 423 | 77 | |
More Information
Set Membership: |
|

COX6B2
Genomic Map