Assay Details
| Gene Symbol: COQ7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014849.1 | NP_001014849.1 | 3 - 4 | 430 | 63 | |
| NM_001113241.1 | NP_001106712.1 | 3 - 4 | 430 | 63 | ||
| GenBank mRNA | BC109578.1 | - | 3 - 4 | 394 | 63 | |
| BT021836.1 | - | 3 - 4 | 448 | 63 | ||
| DT837410.1 | - | 3 - 4 | 431 | 63 | ||
| DT847731.1 | - | 2 - 3 | 164 | 63 | ||
More Information
Set Membership: |
|

COQ7
Genomic Map