Assay Details
| Gene Symbol: PPIL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014869.1 | NP_001014869.1 | 1 - 2 | 60 | 68 | |
| GenBank mRNA | BC102396.1 | - | 1 - 2 | 71 | 68 | |
| BT021028.1 | - | 1 - 2 | 60 | 68 | ||
More Information
Set Membership: |
|

PPIL1
Genomic Map