Assay Details
| Gene Symbol: DUSP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014875.1 | NP_001014875.1 | 5 - 6 | 569 | 65 | |
| GenBank mRNA | BC105553.1 | - | 5 - 6 | 606 | 65 | |
| BT021021.1 | - | 5 - 6 | 569 | 65 | ||
More Information
Set Membership: |
|

DUSP11
Genomic Map