Assay Details
| Gene Symbol: ARL6IP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014891.1 | NP_001014891.1 | 2 - 3 | 400 | 69 | |
| GenBank mRNA | BC102757.1 | - | 2 - 3 | 504 | 69 | |
| BT020899.1 | - | 2 - 3 | 400 | 69 | ||
More Information
Set Membership: |
|

ARL6IP5
Genomic Map