Assay Details
| Gene Symbol: FAM214B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014893.1 | NP_001014893.1 | 3 - 4 | 1197 | 53 | |
| GenBank mRNA | BT021202.1 | - | 3 - 4 | 1197 | 53 | |
More Information
Set Membership: |
|

FAM214B
Genomic Map